Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95010

Sigma-Aldrich

Vinylmagnesium chloride solution

technical, ~1.7 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHMgCl
CAS Number:
Molecular Weight:
86.80
Beilstein/REAXYS Number:
3535268
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical

form

liquid

reaction suitability

reaction type: Grignard Reaction

concentration

~1.7 M in THF

density

0.980 g/mL at 20 °C

SMILES string

Cl[Mg]C=C

InChI

1S/C2H3.ClH.Mg/c1-2;;/h1H,2H2;1H;/q;;+1/p-1

InChI key

NAFZXKWEUWVSAW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

target_organs

Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service