Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53019

Supelco

Eosin Y disodium salt

analytical standard

Synonym(s):

2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, D&C;Red No. 22, Eosin yellowish

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85
Colour Index Number:
45380
Beilstein/REAXYS Number:
3586809
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤4.0% water

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C20H8Br4O5.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

SEACYXSIPDVVMV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Eosin Y disodium salt is a synthetic dye, which is widely used as a photoredox catalyst in organic synthesis.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

contains up to 4% methanol (detected by H-NMR)

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

376.9 °F - Pensky-Martens closed cup

flash_point_c

191.6 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic applications of eosin Y in photoredox catalysis.
Hari PD and Konig B
Chemical Communications (Cambridge, England), 50(51), 6688-6699 (2014)
Hubert Pausch et al.
PLoS genetics, 10(1), e1004044-e1004044 (2014-01-07)
Genetic variants underlying reduced male reproductive performance have been identified in humans and model organisms, most of them compromising semen quality. Occasionally, male fertility is severely compromised although semen analysis remains without any apparent pathological findings (i.e., idiopathic subfertility). Artificial
Jeong-Hae Choi et al.
Archives of dermatological research, 306(7), 635-643 (2014-04-15)
The barrier system of the skin not only defends against antigens and harmful substances, but also hinders the permeation of medicines and cosmetics into the dermis. Several strategies have been developed to enhance the absorption ability of skin, including the
Toshihisa Yashiro et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(48), 17212-17217 (2014-11-19)
Males and females are in conflict over genetic transmission in the evolution of parthenogenesis, because it enhances female reproductive output but deprives the males' genetic contribution. For males, any trait that coerces females into sexual reproduction should increase their fitness.
Mimi C Sammarco et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 29(11), 2336-2345 (2014-04-23)
Amputation of the digit tip within the terminal phalangeal bone of rodents, monkeys, and humans results in near-perfect regeneration of bone and surrounding tissues; however, amputations at a more proximal level fail to produce the same regenerative result. Digit regeneration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service