Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

471968

SAFC

(±)-Propylene oxide

ReagentPlus®, 99%

Synonym(s):

(±)-Methyloxirane, 1,2-Epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Beilstein/REAXYS Number:
79763
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

2 (vs air)

vapor pressure

29.43 psi ( 55 °C)
8.59 psi ( 20 °C)

product line

ReagentPlus®

assay

99%

autoignition temp.

1378 °F

expl. lim.

37 %

refractive index

n20/D 1.366 (lit.)

bp

34 °C (lit.)

mp

−112 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

SMILES string

CC1CO1

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3

InChI key

GOOHAUXETOMSMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-36.4 °F - Equilibrium method

flash_point_c

-38 °C - Equilibrium method

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javix Thomas et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(16), 4582-4587 (2011-03-25)
Chirality recognition in the hydrogen-bonded glycidol···propylene oxide complex has been studied by using rotational spectroscopy and ab initio calculations. An extensive conformational search has been performed for this binary adduct at the MP2/6-311++G(d,p) level of theory and a total of
Sascha Brunke et al.
PLoS pathogens, 10(10), e1004478-e1004478 (2014-10-31)
Candida glabrata is one of the most common causes of candidemia, a life-threatening, systemic fungal infection, and is surpassed in frequency only by Candida albicans. Major factors contributing to the success of this opportunistic pathogen include its ability to readily
Josep Casamada Ribot et al.
Chemical communications (Cambridge, England), 46(37), 6971-6973 (2010-08-24)
The aqueous gelation of a quaternary ammonium oligo(propylene oxide)-based ionic liquid yields ion gels with a reverse thermoresponsive behavior (i.e., mechanical moduli and viscosity increase with temperature) and enhanced ionic conductivities.
Aude Bouchet et al.
The Journal of organic chemistry, 76(10), 4178-4181 (2011-04-08)
ECD and NMR experiments show that the complexation of propylene oxide (PrO) within the cavity of an enantiopure water-soluble cryptophane 1 in NaOH solution is enantioselective and that the (R)-PrO@PP-1 diastereomer is more stable than the (S)-PrO@PP-1 diastereomer with a
Denis Merlet et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 209(2), 315-322 (2011-02-25)
The application of the G-SERFph pulse sequence is presented on enantiomeric mixtures dissolved in a chiral liquid crystal. It aims at editing, within one single 2D spectrum, every proton coupling which is experienced by a given proton site in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service