33731
Trichloroacetic acid
puriss. p.a., ACS reagent, ≥99.5%
Synonym(s):
TCA
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
vapor density
<1 (vs air)
vapor pressure
1 mmHg ( 51 °C)
assay
≥99.5%
impurities
≤0.002% heavy metals (as Pb)
≤0.005% water insolubles
ign. residue
≤0.01% (as SO4)
refractive index
n20/D 1.62 (lit.)
bp
196 °C (lit.)
mp
54-58 °C (lit.)
density
1.62 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤20 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤100 mg/kg
cation traces
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
suitability
complies for appearance of solution
SMILES string
OC(=O)C(Cl)(Cl)Cl
InChI
1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)
InChI key
YNJBWRMUSHSURL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 2
flash_point_f
>235.4 °F - closed cup
flash_point_c
> 113 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service