Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

Skip To

31037

Metolcarb

PESTANAL®, analytical standard

Synonym(s):

m-Tolyl methylcarbamate

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
EC Number:
214-446-0
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
2045865
MDL number:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, NMR: suitable, gas chromatography (GC): suitable

impurities

≤0.5% water (Karl Fischer)

mp

74-77 °C

suitability

passes test for identity (NMR)

format

neat

SMILES string

CNC(=O)Oc1cccc(C)c1

InChI

1S/C9H11NO2/c1-7-4-3-5-8(6-7)12-9(11)10-2/h3-6H,1-2H3,(H,10,11)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Umehara et al.
Journal of neurology, 238(1), 47-48 (1991-02-01)
A 55-year-old woman who attempted suicide by ingesting 200 ml of m-tolyl methyl carbamate (MTMC) is reported. She was comatose for 3 days and, upon recovery, had notable paraesthesia in her lower limbs and difficulty in walking. Neurological examination revealed
Haiyan Liu et al.
Luminescence : the journal of biological and chemical luminescence, 22(4), 302-308 (2007-05-02)
A sensitive, simple and rapid flow-injection chemiluminescence (FI-CL) method is described to determine tsumacide pesticide residue based on the CL reaction of the alkaline degradation product of tsumacide with acidic KMnO(4) when rhodamine 6G was present. Under the optimum conditions
Xiao Liang et al.
Analytica chimica acta, 1050, 139-145 (2019-01-22)
A biosensor that could simultaneously detect multi-analyte as many as in a single test is more favored when facing lots of samples for screening purpose. In such a biosensor, the recognition element with broad specificity and highly affinity is a
Qi Zhang et al.
Analytica chimica acta, 625(1), 87-94 (2008-08-30)
Five different haptens of the N-methylcarbamate insecticide metolcarb were designed and synthesized. All of the haptens were conjugated with ovalbumin (OVA) for the coating antigen, and one hapten containing all of the structure of metolcarb was conjugated with bovine serum
Jia Xia et al.
Analytica chimica acta, 625(1), 28-34 (2008-08-30)
An improved method for the determination of metacrate in water samples has been developed using dispersive liquid-liquid microextraction (DLLME) prior to liquid chromatography analysis. The variables of interest, such as the volume of extraction solvent and disperser solvent, salt effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service