Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M-095

Supelco

Melatonin solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

COc1ccc2[nH]cc(CCNC(C)=O)c2c1

InChI

1S/C13H16N2O2/c1-9(16)14-6-5-10-8-15-13-4-3-11(17-2)7-12(10)13/h3-4,7-8,15H,5-6H2,1-2H3,(H,14,16)

InChI key

DRLFMBDRBRZALE-UHFFFAOYSA-N

General description

Melatonin is a naturally occurring hormone with several effects, notably influencing circadian rhythms. Melatonin is sold as a sleep aid and in certain countries as a dietary supplement. This certified solution standard is suitable for use in melatonin testing applications by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan R Calvo et al.
Journal of pineal research, 55(2), 103-120 (2013-07-31)
Melatonin is the major secretory product synthesized and secreted by the pineal gland and shows both a wide distribution within phylogenetically distant organisms from bacteria to humans and a great functional versatility. In recent years, a considerable amount of experimental
S D Joustra et al.
European journal of endocrinology, 171(2), 217-228 (2014-05-16)
Patients treated for nonfunctioning pituitary macroadenomas (NFMAs) have fatigue and alterations in sleep characteristics and sleep-wake rhythmicity frequently. As NFMAs often compress the optic chiasm, these complaints might be related to dysfunction of the adjacent suprachiasmatic nucleus (SCN). We aimed
Jack Falcón et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 314-319 (2013-12-20)
Melatonin (N-acetyl-5-methoxytrypamine) is the vertebrate hormone of the night: circulating levels at night are markedly higher than day levels. This increase is driven by precisely regulated increases in acetylation of serotonin in the pineal gland by arylalkylamine N-acetyltransferase (AANAT), the
Ane Wilhelmsen-Langeland et al.
Journal of biological rhythms, 28(5), 306-321 (2013-10-18)
Delayed sleep phase disorder (DSPD) is a circadian rhythm sleep disorder. Patients with DSPD have problems initiating sleep if they go to bed at a conventional time, and they often have problems waking at desired times. If they rise early
P Boyce et al.
Acta psychiatrica Scandinavica. Supplementum, (444)(444), 16-23 (2013-08-09)
Disturbances in circadian rhythms have been associated with major depression and may be an underlying mechanism for the disorder. Resynchronisation of circadian rhythms may provide a new approach to treatment, especially by manipulating melatonin secretion. Melatonin is secreted at night

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service