Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

774693

Sigma-Aldrich

7,7-Dichlorobicyclo[4.1.0]heptane

97%

Synonym(s):

7,7-Dichlorobicyclo-[4.1.0]-heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10Cl2
CAS Number:
Molecular Weight:
165.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.503

bp

198 °C/760 mmHg

density

1.208 g/mL at 25 °C

SMILES string

ClC1(Cl)C2CCCCC12

InChI

1S/C7H10Cl2/c8-7(9)5-3-1-2-4-6(5)7/h5-6H,1-4H2

InChI key

MBPSCGRETWPLSI-UHFFFAOYSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

180.0 °F

flash_point_c

82.22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service