Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767662

Sigma-Aldrich

Chloro(1-naphthyl)bis(triphenylphosphine)nickel(II)

97%

Synonym(s):

trans-Chloro(1-naphthyl)bis(triphenylphosphine)nickel(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H37ClNiP2
CAS Number:
Molecular Weight:
745.88
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

reaction suitability

core: nickel
reagent type: catalyst

mp

187-191 °C

storage temp.

20-25°C

SMILES string

Cl[Ni]c1cccc2ccccc12.c3ccc(cc3)P(c4ccccc4)c5ccccc5.c6ccc(cc6)P(c7ccccc7)c8ccccc8

InChI

1S/2C18H15P.C10H7.ClH.Ni/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-6-10-8-4-3-7-9(10)5-1;;/h2*1-15H;1-7H;1H;/q;;;;+1/p-1

InChI key

BFFKUFPCFQLCFH-UHFFFAOYSA-M

Application

Nickel Catalyst for Carbon-Nitrogen Coupling

pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Hua Huang et al.
Organic letters, 13(14), 3750-3753 (2011-06-22)
The amination of triaryl phosphates was achieved using a Ni(II)-(σ-Aryl) complex/NHC catalyst system in dioxane at 110 °C in the presence of NaH as base. Electron-neutral, -rich, and -deficient triaryl phosphates were coupled with a wider range of amine partners

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service