Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632457

Supelco

Methanol solution

suitable for HPLC, contains 10% (v/v) water, 0.10% (v/v) trifluoroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4O
CAS Number:
Molecular Weight:
32.04
Beilstein/REAXYS Number:
3662861
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
technique(s):
HPLC: suitable
bp:
65.4 °C (lit.)
vapor pressure:
410 mmHg

Product Name

Methanol solution, contains 10 % (v/v) water, 0.10 % (v/v) trifluoroacetic acid

vapor density

1.11 (vs air)

Quality Level

vapor pressure

410 mmHg

form

liquid

contains

0.10 % (v/v) trifluoroacetic acid
10 % (v/v) water

expl. lim.

5.5-44 % (lit.)

technique(s)

HPLC: suitable

bp

65.4 °C (lit.)

mp

-98 °C (lit.)

density

0.823 g/mL at 25 °C

format

mixture

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Product filtered through a 0.2 μm filter

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chongwoo Yu et al.
Pharmaceutical research, 19(12), 1907-1914 (2003-01-14)
Resveratrol, a phenolic phytoalexin occurring in grapes, wine, peanuts, and cranberries, has been reported to have anticarcinogenic, antioxidative, phytoestrogenic, and cardioprotective activities. Because little is known about the metabolism of this potentially important compound, the in vitro and in vivo
Nikolai V Ravin et al.
BMC genomics, 14, 837-837 (2013-11-28)
Hansenula polymorpha DL1 is a methylotrophic yeast, widely used in fundamental studies of methanol metabolism, peroxisome biogenesis and function, and also as a microbial cell factory for production of recombinant proteins and metabolic engineering towards the goal of high temperature
K Heinig et al.
Journal of chromatography. B, Biomedical sciences and applications, 735(2), 171-188 (2000-02-12)
Free carnitine and acylcarnitines (carnitine esters) play an important role in the metabolism of fatty acids. Metabolic disorders can be detected by abnormal levels of these compounds in biological fluids. Capillary electrophoresis-mass spectrometry has the advantage of combining an efficient
B S Kaphalia et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 28(2), 264-273 (1995-12-01)
Human exposure to methanol is likely to increase in the future due to its proposed use as an alternate automobile fuel. Since alcohols are known to esterify the fatty acids in the body and some of those esterified esters are
Y S Keum et al.
Cancer letters, 150(1), 41-48 (2001-02-07)
Heat treatment of Panax ginseng C.A. Meyer at a temperature higher than that applied to the conventional preparation of red ginseng yielded a mixture of saponins with potent antioxidative properties. Thus, the methanol extract of heat-processed neoginseng (designated as 'NGMe')

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service