MilliporeSigma
  • Increased endobiotic fatty acid methyl esters following exposure to methanol.
Increased endobiotic fatty acid methyl esters following exposure to methanol.
Fundamental and applied toxicology : official journal of the Society of Toxicology (1995-12-01)
B S Kaphalia, J B Carr, G A Ansari
ABSTRACT
Human exposure to methanol is likely to increase in the future due to its proposed use as an alternate automobile fuel. Since alcohols are known to esterify the fatty acids in the body and some of those esterified esters are toxic, we studied the formation of fatty acid esters of methanol in Long-Evans male rats given a single oral dose of 3.5 g/kg body weight of methanol in saline. Animals given an equal volume of saline served as control. Three rats were euthanized at 1, 3, 6, 12 and 24 hr following the treatment. Fatty acid methyl esters, extracted from whole blood, liver, pancreas, and brown fat were separated by thin-layer chromatography and quantitated by gas chromatography (GC). Their identity was then confirmed by GC-mass spectrometry. Average levels as high as 596, 5293, 2239, 1106, 9665, 7728, 562, and 2792 micrograms/g (wet weight basis) of 14:0, 16:0, 16:1, 18:0, 18:1, 18:2, 18:3, and 20:4 fatty acid methyl esters, respectively, were found in the pancreas of methanol-treated rats. The average concentration of total fatty acid methyl esters was computed to be 4513, 29594, 22871, 18956, 17014, and 9702 micrograms/g in the pancreas compared to 1.9, 25.4, 36.8, 18.5, 18.9, and 14.2 micrograms/g in the liver at 0, 1, 3, 6, 12, and 24 hr, respectively, following methanol exposure. On dry lipid weight basis, the levels were significantly higher again in pancreas followed by brown fat and liver. In whole blood, only low levels of 16:0, 18:0, and 20:4 fatty acid methyl esters could be detected at all time points. The highest concentration of total fatty acid methyl esters in the pancreas, liver, and brown fat was detected at 1, 3, and 24 hr, respectively. Most of the fatty acid methyl esters found in the liver and pancreas decreased after 6 hr of methanol exposure. The fatty acid methyl esters of higher concentrations were 16:0 in the whole blood, 18:0, 18:1, 18:2, and 20:4 in liver, 18:1, and 18:2 in pancreas and 16:0, 18:1, and 18:2 in brown fat. These fatty acid methyl esters were also detected in the tissues of control rats indicating their endogenous formation. Significant increase in methylation of the fatty acids during methanol exposure, as found in this study, may serve as a defense mechanism for preventing available methanol from oxidative metabolism to render toxicity. However, the biological significance of these fatty acid methyl esters is yet to be understood.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%