MilliporeSigma
  • Antioxidant and anti-tumor promoting activities of the methanol extract of heat-processed ginseng.
Antioxidant and anti-tumor promoting activities of the methanol extract of heat-processed ginseng.
Cancer letters (2001-02-07)
Y S Keum, K K Park, J M Lee, K S Chun, J H Park, S K Lee, H Kwon, Y J Surh
ABSTRACT
Heat treatment of Panax ginseng C.A. Meyer at a temperature higher than that applied to the conventional preparation of red ginseng yielded a mixture of saponins with potent antioxidative properties. Thus, the methanol extract of heat-processed neoginseng (designated as 'NGMe') attenuated lipid peroxidation in rat brain homogenates induced by ferric ion or ferric ion plus ascorbic acid. Furthermore, the extract protected against strand scission in phiX174 supercoiled DNA induced by UV photolysis of H2O2, and was also capable of scavenging superoxide generated by xanthine-xanthine oxidase or by 12-O-tetradecanoylphorbol-13-acetate (TPA) in differentiated human promyelocytic leukemia (HL-60) cells. Topical application of NGMe onto shaven backs of female ICR mice 10 min prior to TPA, significantly ameliorated skin papillomagenesis initiated by 7,12-dimethylbenz[a]anthracene. Moreover, TPA-induced enhancement of epidermal ornithine decarboxylase (ODC) activity and ODC mRNA expression was abolished by a topical dose (0.68 mg) of NGMe. Likewise, TPA-induced production of tumor necrosis factor- in mouse skin was inhibited by NGMe pretreatment.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%