Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

379573

Sigma-Aldrich

Iron(III) phthalocyanine chloride

Dye content ~95 %

Synonym(s):

Phthalocyanine iron(III) monochloride salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16ClFeN8
CAS Number:
Molecular Weight:
603.82
Beilstein/REAXYS Number:
1200456
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, ~95%

reaction suitability

reaction type: C-H Activation
reagent type: catalyst

λmax

654 nm

SMILES string

Cl[Fe]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.ClH.Fe/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;;/h1-16H;1H;/q-2;;+3/p-1

InChI key

LFECRQMMUVBGDC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Iron catalyst for highly stereoselective C-H Amination reactions[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shauna M Paradine et al.
Journal of the American Chemical Society, 134(4), 2036-2039 (2012-01-21)
A highly selective C-H amination reaction under iron catalysis has been developed. This novel system, which employs an inexpensive, nontoxic [Fe(III)Pc] catalyst (typically used as an industrial ink additive), displays a strong preference for allylic C-H amination over aziridination and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service