About This Item
Recommended Products
vapor density
3.3 (vs air)
vapor pressure
3380 mmHg ( 20 °C)
assay
99%
expl. lim.
13.5 %
bp
−18-−16 °C (lit.)
SMILES string
FC(F)(F)C=C
InChI
1S/C3H3F3/c1-2-3(4,5)6/h2H,1H2
InChI key
FDMFUZHCIRHGRG-UHFFFAOYSA-N
Packaging
Compatible with the following:
Legal Information
also commonly purchased with this product
hose barb
recommended
regulator
signalword
Danger
hcodes
Hazard Classifications
Flam. Gas 1 - Press. Gas Compr. Gas - STOT RE 2
Storage Class
2A - Gases
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service