MilliporeSigma
  • Competing reaction pathways of 3,3,3-trifluoropropene at rhodium hydrido, silyl and germyl complexes: C-F bond activation versus hydrogermylation.
Competing reaction pathways of 3,3,3-trifluoropropene at rhodium hydrido, silyl and germyl complexes: C-F bond activation versus hydrogermylation.
Dalton transactions (Cambridge, England : 2003) (2016-11-02)
Theresia Ahrens, Michael Teltewskoi, Mike Ahrens, Thomas Braun, Reik Laubenstein
ABSTRACT
The reaction of the silyl complex [Rh{Si(OEt)
Sigma-Aldrich
Tributylgermanium hydride, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylgermanium hydride, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Triphenylgermanium hydride
Pricing and availability is not currently available.