Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

133515

Sigma-Aldrich

2-Aminonorbornane hydrochloride

99%

Synonym(s):

2-Norbornanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13N · HCl
CAS Number:
Molecular Weight:
147.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

mp

295 °C (dec.) (lit.)

SMILES string

Cl.NC1CC2CCC1C2

InChI

1S/C7H13N.ClH/c8-7-4-5-1-2-6(7)3-5;/h5-7H,1-4,8H2;1H/t5-,6+,7?;/m1./s1

InChI key

IZZMPZQAQTXAHW-ZJVNHHPWSA-N

Biochem/physiol Actions

2-Aminonorbornane hydrochloride is a large neutral amino acid transport inhibitor and lowers the brain phenylalanine level in Pahenu2 mice[1].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abstracts of the Annual Symposium of the Society for the Study of Inborn Errors of Metabolism. Birmingham, United Kingdom. September 4-7, 2012.
Journal of inherited metabolic disease, 35 Suppl 1, S1-182 (2012-08-17)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service