MilliporeSigma
  • High expression of CTHRC1 promotes EMT of epithelial ovarian cancer (EOC) and is associated with poor prognosis.
High expression of CTHRC1 promotes EMT of epithelial ovarian cancer (EOC) and is associated with poor prognosis.
Oncotarget (2015-10-10)
Minzhi Hou, Zhiqiang Cheng, Hongwei Shen, Shanyang He, Yang Li, Yunping Pan, Chongjin Feng, Xinlin Chen, Yang Zhang, Millicent Lin, Liantang Wang, Zunfu Ke
ABSTRACT
Collagen triple helix repeat-containing 1 (CTHRC1) is aberrantly overexpressed in multiple malignant tumors. However, the expression characteristics and function of CTHRC1 in epithelial ovarian cancer (EOC) remain unclear. We found that CTHRC1 expression was up-regulated in the paraffin-embedded EOC tissues compared to borderline or benign tumor tissues. CTHRC1 expression was positively correlated with tumor size (p = 0.008), menopause (p = 0.037), clinical stage (p = 0.002) and lymph node metastasis (p < 0.001) and was also an important prognostic factor for the overall survival of EOC patients, as revealed by Kaplan-Meier analysis. CTHRC1 increased the invasive capabilities of EOC cells in vitro by activating the Wnt/β-catenin signaling pathway. We showed that ectopic transfection of CTHRC1 in EOC cells up-regulated the expression of EMT markers such as N-cadherin and vimentin, and EMT-associated transcriptional factor Snail. Knockdown of CTHRC1 expression in EOC cells resulted in down-regulation of N-cadherin, vimentin, Snail and translocation of β-catenin. Collectively, CTHRC1 may promote EOC metastasis through the induction of EMT process and serve as a potential biomarker for prognosis as well as a target for therapy.
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
MISSION® esiRNA, targeting human CTHRC1
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Trioctylphosphine, technical grade, 90%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiourea, JIS special grade, ≥98.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cthrc1
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%