MilliporeSigma
  • In vitro effects of vitamin supplements on platelet-activating factor and its metabolism in age-related macular degeneration.
In vitro effects of vitamin supplements on platelet-activating factor and its metabolism in age-related macular degeneration.
Cutaneous and ocular toxicology (2013-10-24)
Marilita M Moschos, Irini P Chatziralli, George Stamatakis, Vasiliki D Papakonstantinou, Constantinos A Demopoulos
ABSTRACT
The purpose of our study was to investigate for the first time a series of vitamin supplements used for age-related macular degeneration (AMD) as potential inhibitors of platelet-activating factor (PAF). Various vitamin supplements were tested in washed rabbit platelets (WRPs), in order to investigate the interaction between vitamin supplements (InShape, Nutrof, Ocuvite, Vitalux) and inhibition of PAF-induced platelet aggregation. Additionally, we examined their ability to affect PAF-metabolism, through their in vitro effect on PAF basic metabolic enzymes (PAF-CPT, lyso PAF-AT, and PAF-AH). Nutrof exhibited the strongest anti-PAF activity, while Vitalux was the most potent anti-inflammatory factor. This is the first study to bring in surface potent anti-inflammatory and anti-angiogenic activities of some vitamin supplements used against AMD, through their in vitro anti-PAF effects in WRPs and the rabbit plasma and leukocyte PAF metabolism, suggesting a promising role of vitamin supplements and especially resveratrol, concerning its potent anti-angiogenic activity in AMD.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,4-Bis(5-phenyl-2-oxazolyl)benzene, BioReagent, suitable for scintillation
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
SKU
Pack Size
Availability
Price
Quantity
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,5-Diphenyloxazole, suitable for liquid scintillation spectrometry
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,5-Diphenyloxazole, 99%, suitable for scintillation
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Glycerol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture