Skip to Content
MilliporeSigma
Formaldehyde solution 37% (stabilized with about 10% methanol) for synthesis

Formaldehyde solution

Formaldehyde solution 37% (stabilized with about 10% methanol) for synthesis

Products

Product Category
Brand
Physical Form
Product Line
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing