Skip to Content
MilliporeSigma
All Photos(2)

Documents

SRP5049

Sigma-Aldrich

MET (958-end), active, GST tagged from rat

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HGFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

39-53 nmol/min·mg

mol wt

~74 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

rat ... Met(24553)

General description

MET is a proto-oncogene that encodes a transmembrane growth factor receptor which is a heterodimer of two disulphide linked chains of 50 kd (alpha) and 145 kd (beta). MET is widely expressed in the kidney, brain, lung, skin, and embryonic tissue. Hepatocyte growth factor (HGF) binds to MET and activates its tyrosine kinase activity. MET is overexpressed and activated in a variety of human cancers including pancreatic, colon, gastric, cervical and ovarian cancers and has been shown to be involved in tumor cell migration and invasion.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Giordano et al.
Oncogene, 4(11), 1383-1388 (1989-11-01)
The proto-oncogene c-met encodes a transmembrane protein with structural features of a growth factor receptor. We have previously shown that the c-met protein (c-Met) is a heterodimer of two disulphide linked chains of 50 kd (alpha) and 145 kd (beta).
A Iyer et al.
Cell growth & differentiation : the molecular biology journal of the American Association for Cancer Research, 1(2), 87-95 (1990-02-01)
A 6.7-kilobase met complementary DNA (cDNA) was isolated from a pcD cDNA library prepared from C3H mouse fibroblast cell line polyadenylated RNA. Sequence analysis of 6.7-kilobase met cDNA insert revealed that it contained the entire open reading frame and shared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service