Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0707

Sigma-Aldrich

FKGK11

≥98% (GC)

Synonym(s):

1,1,1,2,2-Pentafluoro-7-phenyl-3-Heptanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13F5O
CAS Number:
Molecular Weight:
280.23
UNSPSC Code:
41106300
NACRES:
NA.77

assay

≥98% (GC)

form

oil

color

colorless to yellow

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C13H13F5O/c14-12(15,13(16,17)18)11(19)9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2

InChI key

ROPVXAWEVYWEQY-UHFFFAOYSA-N

Biochem/physiol Actions

FKGK11 is a potent inhibitor of GVIA iPLA2 with little or no inhibition against GIVA cPLA2 (XI(50) = 0.0073 and >0.91, respectively).
FKGK11 is a potent inhibitor of GVIA iPLA2 with little or no inhibition against GIVA cPLA2 (XI(50) = 0.0073 and >0.91, respectively). The compound displays a slight inhibition against GVsPLA2. FKGK11 potently inhibited the progression and severity in a murine experimental autoimmune encephalomyelitis (EAE) model.

Features and Benefits

This compound is featured on the Phospholipase A2 page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Air sensitive; suggest to store under nitrogen in freezer.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Phospholipase A2 (PLA2) enzymes hydrolyze glycerophospholipids, yielding arachidonic acid; inhibition targets eicosanoid biosynthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service