Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0759

Supelco

5α-Pregnane-3α,11β,17α,21-tetrol-20-one

Synonym(s):

3α,11β,17α,21-Tetrahydroxy-5α-pregnan-20-one, 3α,5α-Tetrahydrocortisol, Allopregnane-3α,11β,17α,21-tetrol-20-one, Allotetrahydrocortisol, Kendall’s compound C, Reichstein’s substance C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O5
CAS Number:
Molecular Weight:
366.49
EC Number:
MDL number:
UNSPSC Code:
41116107

assay

≥90%

SMILES string

C[C@@]12CC[C@H](O)C[C@H]1CC[C@@H]3[C@H]4CC[C@@](O)(C(=O)CO)[C@@]4(C)C[C@@H](O)[C@@H]23

InChI

1S/C21H34O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12-16,18,22-24,26H,3-11H2,1-2H3/t12-,13+,14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

AODPIQQILQLWGS-FDSHTENPSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service