Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N4284

Sigma-Aldrich

Neuromedin U-25 human

≥97% (HPLC), lyophilized powder

Synonym(s):

NMU-25

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C141H203N41O38
Molecular Weight:
3080.37
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

assay

≥97% (HPLC)

form

lyophilized powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NMS(129521)

Amino Acid Sequence

Phe-Arg-Val-Asp-Glu-Glu-Phe-Gln-Ser-Pro-Phe-Ala-Ser-Gln-Ser-Arg-Gly-Tyr-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service