Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L5670

Sigma-Aldrich

Locostatin

≥98% (HPLC)

Synonym(s):

(4S)-3-[(E)-1-Oxo-2-butenyl]-4-(phenylmethyl)-2-oxazolidinone, (4S)-3-[(E)-But-2-enoyl]-4-benzyl-2-oxazolidinone, UIC-1005

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$67.80
50 MG
$262.00

$67.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$67.80
50 MG
$262.00

About This Item

Empirical Formula (Hill Notation):
C14H15NO3
CAS Number:
Molecular Weight:
245.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$67.80


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥30 mg/mL

storage temp.

2-8°C

SMILES string

C\C=C\C(=O)N1[C@H](COC1=O)Cc2ccccc2

InChI

1S/C14H15NO3/c1-2-6-13(16)15-12(10-18-14(15)17)9-11-7-4-3-5-8-11/h2-8,12H,9-10H2,1H3/b6-2+/t12-/m0/s1

InChI key

UTZAFVPPWUIPBH-QSLRECBCSA-N

Biochem/physiol Actions

Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration.
Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service