Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C5762

Sigma-Aldrich

Colominic acid sodium salt from Escherichia coli

Synonym(s):

Poly[2,8-(N-acetylneuraminic acid sodium salt)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C11H16NNaO8)n
CAS Number:
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.76

form

powder

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

viruses

mode of action

enzyme | inhibits

storage temp.

−20°C

InChI

1S/C11H20NO7.3Na/c1-6(15)12-10-7(16)4-9(18-3-2-13)19-11(10)8(17)5-14;;;/h3,7-11,13-14,16-17H,2,4-5H2,1H3,(H,12,15);;;/q;3*+1

InChI key

IPYNPWIUCZLARC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Colominic acid is used to study protein polysialylation processes. It is a substrate for CMP-NeuAc:(NeuAcα2→8)n (colominic acid) sialyltransferase.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Roth et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(9), 2999-3003 (1988-05-01)
A unique structural feature of the neural cell adhesion molecule N-CAM is the presence of homopolymers of alpha (2----8)-linked sialic acid units. We have used two specific probes for the detection of poly(sialic acid) in normal human kidney and Wilms
J Finne et al.
Journal of immunology (Baltimore, Md. : 1950), 138(12), 4402-4407 (1987-06-15)
The structurally similar polysialic acid capsules of group B meningococci and Escherichia coli K1 are poor immunogens, and attempts are currently being made to improve their immunogenicity by chemical modifications. An IgG monoclonal antibody to these polysialic acid capsules was
J Roth et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(7), 1969-1973 (1987-04-01)
We report the presence of alpha(2----8)-linked polysialic acid as detected with a monoclonal antibody outside the nervous tissue in the postnatal developing rat kidney. By immunoblot analysis, the reactivity was confined to a broad band of apparent molecular mass 200-250
Nina Westphal et al.
Scientific reports, 7(1), 13631-13631 (2017-10-21)
The neural cell adhesion molecule (NCAM) is the major carrier of polysialic acid (PSA) which modulates NCAM functions of neural cells at the cell surface. In previous studies, we have shown that stimulation of cultured neurons with surrogate NCAM ligands

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Questions

1–2 of 2 Questions  
  1. What is the molecular weight of this polymer?

    1 answer
    1. The molecular weight of this product is not determined. However historical data, as well as various published sources, suggest the molecular weight may range from 10 kDa - 25 kDa.

      Helpful?

  2. What is the molecular weight of this product C5762?

    1 answer
    1. The molecular weight of the product is approximately 30,000 g/mol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service