Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47860

Sigma-Aldrich

Basic Fuchsin

for microscopy (Bact., Bot., Hist.), indicator (pH 1.0-3.1)

Synonym(s):

Basic Violet 14, Benzenamine, 4-[(4-aminophenyl)(4-imino-2,5-cyclohexadien-1-ylidene)methyl]-2-methyl hydrochloride, Benzenamine, 4-[(4-aminophenyl)(4-imino-2,5-cyclohexadien-1-ylidene)methyl]-2-methyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN3
CAS Number:
Molecular Weight:
337.85
Colour Index Number:
42510
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bact., Bot., Hist.)
indicator (pH 1.0-3.1)

application(s)

environmental

SMILES string

NC1=CC=C(C(C2=CC(C)=C(N)C=C2)=C3C=CC(C=C3)=N)C=C1.[Cl]

InChI

1S/C20H19N3.ClH/c1-13-12-16(6-11-19(13)23)20(14-2-7-17(21)8-3-14)15-4-9-18(22)10-5-15;/h2-12,23H,21-22H2,1H3;1H

InChI key

AXDJCCTWPBKUKL-UHFFFAOYSA-N

General description

Basic fuchsin, a triphenylmethane dye is commonly a mixture of pararosaniline (C.I. 42500), rosaniline (C.I. 42510), magenta II (no CI number) and new fuchsine (CI 42520). Basic fuchsin is widely used in tissue-specific staining of biological and coloring several materials including orlon, cotton, leather, paper etc. It is also used to stain collagen, muscle and mitochondria.

Application

Basic Fuchsin has been used in several biological tests including:
  • Acid-fast staining with carbol-fuchsine
  • Periodic Acid-Schiff method
  • Feulgen technique for DNA staining
  • ENDO culture medium for distinguishing between the bacteria Escherichia coli and Aerobacter aerogenes
  • Leifson′s method for bacterial flagella staining.

Biochem/physiol Actions

Basic fuchsin possesses anesthetic, bactericidal (gram-positive), and fungicidal properties.

Analysis Note

homologous mixture of dyes, contains dominantly (~35-40%) Basic Fuchsin (C.I. 42510)

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daria Bloch et al.
Development (Cambridge, England), 146(17) (2019-08-11)
Patterning of the root xylem into protoxylem (PX) and metaxylem is regulated by auxin-cytokinin signaling and microRNA miR165a/166b-mediated suppression of genes encoding Class III HOMEODOMAIN LEU-ZIPPER (HD-ZIPIII) proteins. We found that, in Arabidopsis, osmotic stress via core abscisic acid (ABA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service