Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37687

Supelco

1,4-Dithiane-2,5-diol

technical, ≥90% (S)

Synonym(s):

2,5-Dihydroxy-1,4-dithiane, Mercaptoacetaldehyde dimer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2S2
CAS Number:
Molecular Weight:
152.24
Beilstein/REAXYS Number:
104402
MDL number:
UNSPSC Code:
41113319
PubChem Substance ID:

grade

technical

Quality Level

assay

≥90% (S)

mp

130 °C (dec.) (lit.)

application(s)

general analytical

storage temp.

2-8°C

SMILES string

OC1CSC(O)CS1

InChI

1S/C4H8O2S2/c5-3-1-7-4(6)2-8-3/h3-6H,1-2H2

InChI key

YUIOPHXTILULQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Inamori et al.
Chemical & pharmaceutical bulletin, 38(1), 243-245 (1990-01-01)
2,5-Dihydroxy-1,4-dithiane (I) and its derivatives (II-IV) showed rather marked inhibitory activities on the growth of the roots of two plant species. All compounds tested had phytogrowth-inhibitory activities. These compounds markedly inhibited the growth of the two plant species at the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service