Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

183954

Sigma-Aldrich

1,4-Dithiane-2,5-diol

97%

Synonym(s):

2,5-Dihydroxy-1,4-dithiane, Mercaptoacetaldehyde dimer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2S2
CAS Number:
Molecular Weight:
152.24
Beilstein/REAXYS Number:
104402
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

mp

130 °C (dec.) (lit.)

functional group

hydroxyl
thioether

storage temp.

2-8°C

SMILES string

OC1CSC(O)CS1

InChI

1S/C4H8O2S2/c5-3-1-7-4(6)2-8-3/h3-6H,1-2H2

InChI key

YUIOPHXTILULQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dithiane-2,5-diol undergoes bifunctional squaramide catalyzed sulfa-Michael/aldol cascade reaction with chalcones to afford trisubstituted tetrahydrothiophenes[1]. It participates in diastereoselective [3+3] cycloaddition with azomethine imines catalyzed by 1,4-diazabicyclo[2.2.2]octane[2]. It is a mercaptoacetaldehyde dimer and an efficient synthon for incorporating a thiol group to in situ generated nitroalkenes[3].

Application

1,4-Dithiane-2,5-diol was used in the synthesis of:
  • substituted tetrahydrothiophene derivatives[3]
  • 2-amino-3-(arylsulfonyl)thiophenes, potential antiviral and antitumor agents[4]

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1A - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 4-Dithiane-2, 5-diol as an efficient synthon for a straightforward synthesis of functionalized tetrahydrothiophenes via sulfa-Michael/aldol-type reactions with electrophilic alkenes.
Baricordi N, et al.
Tetrahedron, 68(1), 208-213 (2012)
Xin Fang et al.
Organic letters, 15(21), 5554-5557 (2013-11-05)
An effective diastereoselective [3 + 3] cycloaddition of 1,4-dithiane-2,5-diol with azomethine imines catalyzed by DABCO is described. A variety of highly functionalized six-membered dinitrogen-fused heterocycles can be obtained in good yield with excellent diastereoselectivity, which was controlled by anomeric effect.
C E Stephens et al.
Bioorganic & medicinal chemistry, 9(5), 1123-1132 (2001-05-30)
Based on general SARs previously described for anti-HIV-1 diarylsulfone derivatives, a series of 2-amino- and 2-carboxamido-3-arylsulfonylthiophenes has been prepared and evaluated as potential antiviral and antitumor agents. In cell culture, some of the 2-aminothiophenes exhibited moderate and selective activity against
Jun-Bing Ling et al.
Organic letters, 14(4), 1090-1093 (2012-02-09)
A bifunctional squaramide catalyzed sulfa-Michael/aldol cascade reaction between 1,4-dithiane-2,5-diol and chalcones with a low catalyst loading has been developed. Trisubstituted tetrahydrothiophenes with three contiguous stereogenic centers are obtained in a highly stereocontrolled manner. Additionally, a remarkable temperature effect on reaction
Y Inamori et al.
Chemical & pharmaceutical bulletin, 38(1), 243-245 (1990-01-01)
2,5-Dihydroxy-1,4-dithiane (I) and its derivatives (II-IV) showed rather marked inhibitory activities on the growth of the roots of two plant species. All compounds tested had phytogrowth-inhibitory activities. These compounds markedly inhibited the growth of the two plant species at the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service