49904
Ponceau 3R
analytical standard
Synonym(s):
Disodium 3-hydroxy-4-[(2,4,5-trimethylphenyl)azo]-2,7-naphthalenedisulfonate
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
[Na+].[Na+].Cc1cc(C)c(cc1C)\N=N\c2c(O)c(cc3cc(ccc23)S([O-])(=O)=O)S([O-])(=O)=O
InChI
1S/C19H18N2O7S2.2Na/c1-10-6-12(3)16(7-11(10)2)20-21-18-15-5-4-14(29(23,24)25)8-13(15)9-17(19(18)22)30(26,27)28;;/h4-9,22H,1-3H3,(H,23,24,25)(H,26,27,28);;/q;2*+1/p-2/b21-20+;;
InChI key
SWPPXVFKWWEYSV-SERMZQFOSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Recommended products
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service