Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42073

Supelco

13-cis-4-Oxoretinoic acid

analytical standard

Synonym(s):

13-cis-4-oxo-RA, 4-Oxoisotretinoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26O3
CAS Number:
Molecular Weight:
314.42
Beilstein/REAXYS Number:
6067353
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

CC(/C=C/C1=C(C)C(CCC1(C)C)=O)=C\C=C\C(C)=C(C(O)=O)\[H]

InChI

1S/C20H26O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7+,15-13-

Inchi Key

GGCUJPCCTQNTJF-FAOQNJJDSA-N

General description

13-cis-4-Oxoretinoic acid, a polar retinoid metabolite that is structurally related to vitamin A, is vital for vision, growth, reproduction and various other physiological functions.[1]

Application

13-cis-4-Oxoretinoic acid may be used as an analytical standard for the determination of the analyte in biological samples by high performance liquid chromatography (HPLC).[2][3][4][5]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Lanvers et al.
Journal of chromatography. B, Biomedical applications, 685(2), 233-240 (1996-10-25)
All-trans-retinoic acid (all-trans-RA) and 13-cis-retinoic acid (13-cis-RA), due to their effects on cell differentiation, proliferation and angiogenesis, improved treatment results in some malignancies. Pharmacokinetic studies of all-trans-RA and 13-cis-RA along with monitoring of retinoic acid metabolites may help to optimize
C Eckhoff et al.
Biomedical & environmental mass spectrometry, 19(7), 428-433 (1990-07-01)
Eight metabolites of retinol were isolated by high-performance liquid chromatography (HPLC) from the plasma of the non-human primate Macaca fascicularis after acute exposure to 150,000 IU of vitamin A per kilogram body weight. After enrichment and further chromatographic purification, the
C Eckhoff et al.
Journal of lipid research, 31(8), 1445-1454 (1990-08-01)
Human plasma was analyzed by high performance liquid chromatography for the presence of retinoic acid and 4-oxoretinoic acid isomers. Peaks that coeluted with the reference compounds all-trans-retinoic acid, 13-cis-retinoic acid, and 13-cis-4-oxoretinoic acid were routinely observed in human plasma. These
T Teerlink et al.
Journal of chromatography. B, Biomedical sciences and applications, 694(1), 83-92 (1997-06-20)
A reversed-phase high-performance liquid chromatographic method for the simultaneous analysis of retinol, all-trans-retinoic acid, 13-cis-retinoic acid and 13-cis-4-oxoretinoic acid in human plasma and cell culture medium is described. Sample preparation involves precipitation of proteins and extraction of retinoids with 60%
Carsten K Schmidt et al.
Analytical biochemistry, 315(1), 36-48 (2003-04-04)
We present a reliable, highly sensitive, and versatile method for the simultaneous determination of endogenous polar (acidic) and apolar (retinol, retinal, and retinyl esters) retinoids in various biological matrices. Following a single liquid extraction of retinoids from tissues or plasma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service