Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32340

Supelco

Tris(2,3-dichloropropyl) phosphate

analytical standard

Synonym(s):

Phosphoric acid tris(2,3-dichloropropyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15Cl6O4P
CAS Number:
Molecular Weight:
430.90
Beilstein/REAXYS Number:
1715433
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

O=P(OCC(Cl)CCl)(OCC(Cl)CCl)OCC(CCl)Cl

InChI

1S/C9H15Cl6O4P/c10-1-7(13)4-17-20(16,18-5-8(14)2-11)19-6-9(15)3-12/h7-9H,1-6H2

InChI key

JZZBTMVTLBHJHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R K Lynn et al.
Drug metabolism and disposition: the biological fate of chemicals, 9(5), 434-441 (1981-09-01)
The metabolism, distribution, and excretion of the flame retardant, tris(1,3-dichloro-2-propyl) phosphate (TDCP), were studied in the rat. Five days after iv administration of 14C-TDCP, 92% of the administered radiolabel had been excreted in the urine (54%), feces (16%), and expired
T Hudec et al.
Science (New York, N.Y.), 211(4485), 951-952 (1981-02-27)
Negative-chemical-ionization mass spectral screening of extracts of human seminal plasma has revealed a presence of a Cl7 ion cluster at a mass-to-charge ratio (m/z) of 463 in a significant number of the samples examined (34 out of 123). Experiments with
The effects of orthophenylphenol, tris(2,3-dichloropropyl) phosphate, and cyclophosphamide on the immune system and host susceptibility of mice following subchronic exposure.
M I Luster et al.
Toxicology and applied pharmacology, 58(2), 252-261 (1981-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service