Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21497

Sigma-Aldrich

4-Nitrophenyl decanoate

≥99.0% (TLC)

Synonym(s):

4-Nitrophenyl caprate, Decanoic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23NO4
CAS Number:
Molecular Weight:
293.36
Beilstein/REAXYS Number:
1915230
MDL number:
UNSPSC Code:
12352204

assay

≥99.0% (TLC)

mp

~35 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C16H23NO4/c1-2-3-4-5-6-7-8-9-16(18)21-15-12-10-14(11-13-15)17(19)20/h10-13H,2-9H2,1H3

InChI key

RRNKCSIEGPOIKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service