Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08490

Sigma-Aldrich

2-Amino-5-methylhexane

≥99.0% (GC)

Synonym(s):

1,4-Dimethylpentylamine, 5-Methyl-2-hexylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2CH(CH3)NH2
CAS Number:
Molecular Weight:
115.22
Beilstein/REAXYS Number:
1731716
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Product 08490 is not currently sold in your country. Contact Technical Service

Quality Level

assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.416

bp

128-129 °C (lit.)

density

0.760 g/mL at 20 °C (lit.)

SMILES string

N[C@H](C)CCC(C)C

InChI

1S/C7H17N/c1-6(2)4-5-7(3)8/h6-7H,4-5,8H2,1-3H3

InChI key

IZCBXLKODYZSDJ-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

82.4 °F - closed cup

flash_point_c

28 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Li et al.
Analytical chemistry insights, 7, 47-58 (2012-08-24)
A sensitive and reliable method of liquid chromatography-electrospray ionization/tandem mass spectrometry (LC-ESI/MS/ MS) was developed and validated for determining 1,3-dimethylamylamine (1,3-DMAA) and 1,4-dimethylamylamine (1,4-DMAA) in geranium plants (Pelargonium graveolens). The sample was extracted with 0.5 M HCl and purified by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service