MilliporeSigma
  • Identification and quantification of dimethylamylamine in geranium by liquid chromatography tandem mass spectrometry.
Identification and quantification of dimethylamylamine in geranium by liquid chromatography tandem mass spectrometry.
Analytical chemistry insights (2012-08-24)
J S Li, M Chen, Z C Li
ABSTRACT
A sensitive and reliable method of liquid chromatography-electrospray ionization/tandem mass spectrometry (LC-ESI/MS/ MS) was developed and validated for determining 1,3-dimethylamylamine (1,3-DMAA) and 1,4-dimethylamylamine (1,4-DMAA) in geranium plants (Pelargonium graveolens). The sample was extracted with 0.5 M HCl and purified by liquid-liquid partition with hexane. The parameters for reverse-phase (C18) LC and positive ESI/MS/MS were optimized. The matrix effect, specificity, linearity, precision, accuracy and reproducibility of the method were determined and evaluated. The method was linear over a range of 0.10-10.00 ng/mL examined, with R(2) of 0.99 for both 1,3-DMAA and 1,4-DMAA. The recoveries from spiked concentrations between 5.00-40.00 ng/g were 85.1%-104.9% for 1,3-DMAA, with relative standard deviation (RSD) of 2.9%-11.0%, and 82.9%-101.8% for 1,4-DMAA, with RSD of 3.2%-11.7%. The instrument detection limit was 1-2 pg for both DMAAs. The quantification limit was estimated to be 1-2 ng/g for the plant sample. This method was successfully applied to the quantitative determination of 1,3- and 1,4-DMAA in both geranium plant and geranium oil.