Key Documents
19-050A
Omni International
Omni Bead Ruptor 12
About This Item
PDF Icon
Shipped & Sold By Omni International
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Omni International
Related Categories
General description
The Bead Ruptor 12 is the most powerful and economical bead mill homogenizer available today. It is an ideal solution for a variety of applications that require grinding, lysing, and homogenization of biological samples prior to molecular extraction.
Using disposable sample tubes pre-filled with a variety of lysing beads, the Bead Ruptor 12 vigorously and uniformly shakes up to 16 tubes at a time providing an efficient, consistent homogenization and high-yield. Unlike most other bead mill homogenizers, the Bead Ruptor 12 does not require a cool down period between runs. This means more samples can be processed in a smaller amount of time.
Benefits of the Bead Ruptor 12 Homogenizer include:
  • Simultaneously homogenize up to 12 x 0.5 or 2 mL tubes and 4 x 7 mL tubes
  • Convenient front-loading tube holder position for ease of use and optimal bead/sample interactions
  • Processing range is ideal for soft tissue samples
  • No cool-down required between runs = non-stop processing
  • Disposable tubes ensure no threat of cross-contamination
  • No sample degradation due to quick processing
  • Features and integrated lid lock
Choose from one of the most recent versions:
Certificates of Analysis (COA)