Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AG777

Sigma-Aldrich

Aquaporin 4, control peptide for AB3068

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$274.00

$274.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$274.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.77

$274.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

biological source

rat

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

inhibition assay: suitable (peptide)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

rat ... AQP4(25293)

General description

17 amino acid peptide sequence from Rat AQP4 from the C-terminus (1,2).

Application

For use in blocking the reactivity of AB3068.

Optimal working dilution must be determined by the end user.

Physical form

Liquid in PBS, pH 7.5 containing 0.05% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Hasegawa et al.
The Journal of biological chemistry, 269(8), 5497-5500 (1994-02-25)
Two mercurial-inhibitable water-transporting proteins have been identified: CHIP28, an erythrocyte water channel also expressed in kidney tubules and selected extrarenal epithelia, and WCH-CD, a kidney collecting duct water channel. In searching for a protein responsible for the high transcellular water

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service