Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-067

Supelco

Albuterol solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21NO3
CAS Number:
Molecular Weight:
239.31
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

concentration

1.0 mg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

CC(C)(C)NCC(O)c1ccc(O)c(CO)c1

General description

Albuterol, or Salbutamol, is a bronchodilator used for the treatment on asthma. This Certified Snap-N-Spike® Solution is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research. Albuterol is marketed under trade names Ventolin®, Athalin, Proventil®, and ProAir®.

Legal Information

ProAir is a registered trademark of Teva Respiratory, LLC
Proventil is a registered trademark of MSD
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Ventolin is a registered trademark of Glaxo Group Limited

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberta Pisi et al.
Allergy and asthma proceedings, 34(1), e14-e20 (2013-02-15)
Small airways are relevant to the pathophysiology of asthma. We investigated whether in asthmatic patients with normal forced expiratory volume in the 1st second (FEV(1)) values, impulse oscillometry system (IOS), as a measure of small airway function, contributed additional information
Diego Gatta et al.
International journal of chronic obstructive pulmonary disease, 8, 169-173 (2013-04-10)
Baseline high neuromuscular drive is present in chronic obstructive pulmonary disease (COPD). In moderate-to-very severe COPD patients, both static and/or dynamic pulmonary hyperinflation have been demonstrated at rest. To assess the influence of dynamic hyperinflation on neuromuscular drive at rest.
Gregory J Feldman
International journal of chronic obstructive pulmonary disease, 8, 89-96 (2013-02-23)
Chronic obstructive pulmonary disease (COPD) is a common disease in the general population and it places a considerable burden on patients, with the disease negatively affecting quality of life. In practice, patients with COPD generally seek medical attention because of
Moyu Ming et al.
Immunotherapy, 5(2), 191-197 (2013-02-19)
Bacillus Calmette-Guérin and other mycobacterial vaccines are important therapeutic methods in a series of chronic inflammatory disorders characterized by Th1/Th2 imbalance in which Th2 type cells and cytokines often increase. However, few studies have investigated whether it can reduce or
Abebaw M Yohannes et al.
International journal of chronic obstructive pulmonary disease, 8, 117-125 (2013-03-22)
Tiotropium bromide is an anticholinergic agent that has gained worldwide acceptance as a first-line, once daily maintenance therapy for patients with moderate-to-severe chronic obstructive pulmonary disease. The purpose of this review is to synthesize the evidence base in the past

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service