Key Documents
Z117420
Sigma-Aldrich
Jacketed addition funnel with pressure-equalization arm
capacity 250 mL
Synonym(s):
funnel glass, glass funnel, glass funnels
Select a Size
1 EA
$1,410.00
$1,410.00
Available to ship onApril 22, 2025Details
Select a Size
Change View
1 EA
$1,410.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
$1,410.00
Available to ship onApril 22, 2025Details
capacity
250 mL
Related Categories
General description
PTFE stopcock and ST/NS: 24/40 joints.
Choose from one of the most recent versions:
Certificates of Analysis (COA)