Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W316350

Sigma-Aldrich

2-Furyl methyl ketone

natural, ≥99%

Synonym(s):

2-Acetylfuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
FEMA Number:
3163
Beilstein/REAXYS Number:
107909
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.054

grade

Kosher
natural

assay

≥99%

refractive index

n20/D 1.5070 (lit.)

bp

67 °C/10 mmHg (lit.)

mp

26-28 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccco1

InChI

1S/C6H6O2/c1-5(7)6-3-2-4-8-6/h2-4H,1H3

InChI key

IEMMBWWQXVXBEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural Occurrence: Burley tobacco, chestnuts, cocoa, coffee, cooked beef and pork, raisins, roasted almonds and peanuts. tamarind, tea, yogurt, wheat bread.

Biochem/physiol Actions

Taste at 10 ppm

comparable product

Product No.
Description
Pricing

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 76-76 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service