Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T77402

Sigma-Aldrich

2,3,5-Trimethylnaphthalene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H5(CH3)3
CAS Number:
Molecular Weight:
170.25
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

refractive index

n20/D 1.608 (lit.)

bp

285 °C/762 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

SMILES string

Cc1cc2cccc(C)c2cc1C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sangwoo Lee et al.
Environmental science. Processes & impacts, 19(9), 1117-1125 (2017-08-08)
Polycyclic aromatic hydrocarbons (PAHs) and alkylated PAHs are known to be major toxic contaminants in spills of petroleum hydrocarbons (oil). Spilled oil undergoes weathering and over time, PAHs go through a series of compositional changes. PAHs can disrupt endocrine functions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service