N19907
4-Nitrophenyl acetate
97%
Synonym(s):
Acetic acid 4-nitrophenyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CO2C6H4NO2
CAS Number:
Molecular Weight:
181.15
Beilstein/REAXYS Number:
515874
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
assay
97%
mp
75-77 °C (lit.)
SMILES string
CC(=O)Oc1ccc(cc1)[N+]([O-])=O
InChI
1S/C8H7NO4/c1-6(10)13-8-4-2-7(3-5-8)9(11)12/h2-5H,1H3
Inchi Key
QAUUDNIGJSLPSX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mirna Velki et al.
Aquatic toxicology (Amsterdam, Netherlands), 193, 187-200 (2017-11-03)
The zebrafish as a test organism enables the investigation of effects on a wide range of biological levels from molecular level to the whole-organism level. The use of fish embryos represents an attractive model for studies aimed at understanding toxic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service