Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A54407

Sigma-Aldrich

2-Aminoethyl hydrogen sulfate

97%

Synonym(s):

Sulfuric acid mono 2-aminoethylester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OSO3H
CAS Number:
Molecular Weight:
141.15
Beilstein/REAXYS Number:
1704079
EC Number:
MDL number:
UNSPSC Code:
12352100

assay

97%

mp

277 °C (dec.) (lit.)

SMILES string

NCCOS(O)(=O)=O

InChI

1S/C2H7NO4S/c3-1-2-7-8(4,5)6/h1-3H2,(H,4,5,6)

InChI key

WSYUEVRAMDSJKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Herbison et al.
Journal of neurochemistry, 55(5), 1617-1623 (1990-11-01)
The characteristics of gamma-aminobutyric acid (GABA) release as monitored by microdialysis have been investigated in the chloral hydrate anaesthetised rat. The high outflow of GABA following insertion of the microdialysis probe (membrane 2 mm in length, 0.5 mm in diameter)
H Golan et al.
Journal of neurophysiology, 71(1), 48-58 (1994-01-01)
1. The cytosolic concentration of a neurotransmitter is believed to be an important factor determining its release. The effects of ethanolamine-O-sulfate (EOS), a gamma-aminobutyric acid (GABA)-transaminase blocker, on GABAergic postsynaptic and presynaptic inhibitory neurotransmission were examined in the crayfish opener
C C Sykes et al.
Journal of neurochemistry, 46(1), 213-217 (1986-01-01)
Ethanolamine O-sulphate (400 mg/kg, i.p.) was administered to rat pups at 9 days of age and on alternate days up to 17 days of age. At 18 days of age, gamma-aminobutyric acid (GABA) concentration was increased (three- to fourfold), glutamic
J P Loeffler et al.
Neuroendocrinology, 43(4), 504-510 (1986-01-01)
The inhibitory action of gamma-aminobutyric acid (GABA) on prolactin (PRL) messenger ribonucleic acid (mRNA) levels was studied in vitro in rat anterior pituitary cells in culture and in intact rats in vivo. PRL mRNA levels were determined by hybridization of
M Qume et al.
Biochemical pharmacology, 52(9), 1355-1363 (1996-11-08)
The inhibitory neurotransmitter gamma-aminobutyric acid (GABA) is not solely located in the CNS, it and the enzymes responsible for its synthesis (glutamic acid decarboxylase, GAD, EC 4.1.1.15) and catabolism (GABA-transaminase, GABA-T, EC 2.6.1.19) are also present in non-neuronal organs. Following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service