779261
2-Azidoanisole solution
0.5 M in tert-butyl methyl ether, ≥95% (HPLC)
Synonym(s):
1-Azido-2-methoxybenzene solution, 2-Methoxyphenyl azide
About This Item
Recommended Products
assay
≥95% (HPLC)
form
liquid
concentration
0.5 M in tert-butyl methyl ether
impurities
≤2.0% water
storage temp.
−20°C
SMILES string
COc1ccccc1N=[N+]=[N-]
InChI
1S/C7H7N3O/c1-11-7-5-3-2-4-6(7)9-10-8/h2-5H,1H3
InChI key
XBXJQBGKICADKR-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-18.4 °F
flash_point_c
-28 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service