Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

766550

Sigma-Aldrich

2,6-Bis(trimethylstannyl)-4,8-bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b′]dithiophene

99.5% (HPLC)

Synonym(s):

1,1′-[4,8-Bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl]bis[1,1,1-trimethyl]stannane, 2,6-Bis(trimethyltin)-4,8-bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b′]dithiophene, 4,8-Bis(2-ethylhexyloxy)-2,6-bis(trimethylstannanyl)-4,8-dihydro-1,5-dithia-s-indacene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H54O2S2Sn2
CAS Number:
Molecular Weight:
772.32
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

99.5% (HPLC)

form

solid

mp

70-75 °C

InChI

1S/C26H36O2S2.6CH3.2Sn/c1-5-9-11-19(7-3)17-27-23-21-13-15-30-26(21)24(22-14-16-29-25(22)23)28-18-20(8-4)12-10-6-2;;;;;;;;/h13-14,19-20H,5-12,17-18H2,1-4H3;6*1H3;;

InChI key

XXMOZDBOAIICDA-UHFFFAOYSA-N

Application

Synthesis of polymers for high power conversion efficiency (PCE) organic solar cells and OFETs

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hae Jung Son et al.
Journal of the American Chemical Society, 133(6), 1885-1894 (2011-01-27)
Herein, we describe the synthesis of fluorinated polythienothiophene-co-benzodithiophenes (PTBFs) and the characterization of their physical properties, especially their performance in solar cells. Fluorination of the polymer backbone lowered both the HOMO and LUMO energy levels and simultaneously widened the energy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service