Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740152

Sigma-Aldrich

2-Hydroxypyrazine

97%

Synonym(s):

2-(1H)-Pyrazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O
CAS Number:
Molecular Weight:
96.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

mp

185-190 °C

SMILES string

Oc1cnccn1

InChI

1S/C4H4N2O/c7-4-3-5-1-2-6-4/h1-3H,(H,6,7)

InChI key

HUTNOYOBQPAKIA-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiajia Mou et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 16(3), 307-325 (2019-06-27)
The cell cycle is regulated by cyclin-dependent kinases (CDKs) and their cognate cyclins, along with their endogenous inhibitors (CDKIs). CDKs act as central regulators in this process. Different CDKs play relevant roles in different phases. Among all CDKs, CDK1 is
Katherine E Frato
Journal of agricultural and food chemistry, 67(1), 341-351 (2018-12-14)
The goal of this study is to identify Coffea arabica O-methyltransferase (OMT) genes involved in the biosynthesis of methoxypyrazines. High levels of 2-isopropyl-3-methoxypyrazine (IPMP) and 2-isobutyl-3-methoxypyrazine (IBMP) in coffee beans are associated with the potato taste defect (PTD). Among the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service