Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

716804

Sigma-Aldrich

Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H36Cl2N2P2Ru
CAS Number:
Molecular Weight:
658.54
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

290 °C (dec.)

SMILES string

Cl[Ru]Cl.NCCCP(c1ccccc1)c2ccccc2.NCCCP(c3ccccc3)c4ccccc4

InChI

1S/2C15H18NP.2ClH.Ru/c2*16-12-7-13-17(14-8-3-1-4-9-14)15-10-5-2-6-11-15;;;/h2*1-6,8-11H,7,12-13,16H2;2*1H;/q;;;;+2/p-2

InChI key

MLZFSEGMJUGESY-UHFFFAOYSA-L

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service