Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

685488

Sigma-Aldrich

1H-Pyrrolo[2,3-f]quinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2
CAS Number:
Molecular Weight:
168.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

235-240 °C

SMILES string

c1cnc2ccc3cc[nH]c3c2c1

InChI

1S/C11H8N2/c1-2-9-10(12-6-1)4-3-8-5-7-13-11(8)9/h1-7,13H

InChI key

IVCGJOSPVGENCT-UHFFFAOYSA-N

Application

Useful fused pyridino-indole for pharmaceutical research.[1]

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason Siu et al.
Organic & biomolecular chemistry, 2(2), 160-167 (2004-01-23)
The development of enhanced conditions for Lewis acid catalysed Leimgruber-Batcho indole synthesis using microwave acceleration is described. This approach has permitted the preparation of a variety of heteroaromatic enamine intermediates in good yield and high purities. Subsequent catalytic hydrogenation reactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service