Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682934

Sigma-Aldrich

2,2′-Dibromobenzophenone

≥90.0%

Synonym(s):

Bis(2-bromophenyl)methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8Br2O
CAS Number:
Molecular Weight:
340.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥90.0%

form

solid

mp

83-87 °C

SMILES string

Brc1ccccc1C(=O)c2ccccc2Br

InChI

1S/C13H8Br2O/c14-11-7-3-1-5-9(11)13(16)10-6-2-4-8-12(10)15/h1-8H

InChI key

JCVYQVHAGOZGIX-UHFFFAOYSA-N

pictograms

Environment

hcodes

pcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service