Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667137

Sigma-Aldrich

5-Bromo-3-pyridinecarboxamide

97%

Synonym(s):

5-Bromonicotinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BrN2O
CAS Number:
Molecular Weight:
201.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

219-223 °C

functional group

amide
bromo

SMILES string

NC(=O)c1cncc(Br)c1

InChI

1S/C6H5BrN2O/c7-5-1-4(6(8)10)2-9-3-5/h1-3H,(H2,8,10)

Inchi Key

YOQRXZIMSKLRCY-UHFFFAOYSA-N

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael A Seidel et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 974, 83-89 (2014-12-03)
Bilirubin oxidation end products (BOXes) appear upon endogenous heme degradation and can be found in the cerebrospinal fluid after hemorrhagic stroke. BOXes are assumed to contribute to delayed cerebral vasospasm and secondary loss of brain tissue. Here, we present a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service