MilliporeSigma
  • Simultaneous determination of the bilirubin oxidation end products Z-BOX A and Z-BOX B in human serum using liquid chromatography coupled to tandem mass spectrometry.
Simultaneous determination of the bilirubin oxidation end products Z-BOX A and Z-BOX B in human serum using liquid chromatography coupled to tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
Raphael A Seidel, Marcel Kahnes, Michael Bauer, Georg Pohnert
ABSTRACT
Bilirubin oxidation end products (BOXes) appear upon endogenous heme degradation and can be found in the cerebrospinal fluid after hemorrhagic stroke. BOXes are assumed to contribute to delayed cerebral vasospasm and secondary loss of brain tissue. Here, we present a validated LC-ESI-MS/MS method for the sensitive determination of the regio-isomers Z-BOX A and Z-BOX B in human serum. We found that Z-BOX A and Z-BOX B appear in serum of healthy volunteers. The sample preparation includes the addition of 5-bromonicotinamide as internal standard and protein precipitation with acetonitrile. Baseline-separation was achieved on a C-18 column with a binary solvent gradient of formic acid in water/acetonitrile at 1 mL/min within a total analysis time of 17 min. Using single reaction monitoring in the positive ion mode, the linear working ranges were 2.74-163 pg/μL (Z-BOX A) and 2.12-162.4 pg/μL (Z-BOX B) with R(2)>0.995. Intra- and inter-day precisions were <10%. The inherent analyte concentrations of Z-BOX A (14.4 ± 5.1 nM) and Z-BOX B (10.9 ± 3.1 nM) in pooled human serum were determined by standard addition. The photolability of both analytes was demonstrated. This method enables to monitor Z-BOX A and Z-BOX B as a prerequisite to systematically study the biological significance of higher order metabolites of heme degradation.
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%