Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

494127

Sigma-Aldrich

Benzyl 3-pyrroline-1-carboxylate

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO2
CAS Number:
Molecular Weight:
203.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

90%

refractive index

n20/D 1.544 (lit.)

bp

133-137 °C/0.6 mmHg (lit.)

density

1.132 g/mL at 25 °C (lit.)

SMILES string

O=C(OCc1ccccc1)N2CC=CC2

InChI

1S/C12H13NO2/c14-12(13-8-4-5-9-13)15-10-11-6-2-1-3-7-11/h1-7H,8-10H2

InChI key

XSKKIFJNZPNVGO-UHFFFAOYSA-N

General description

Benzyl 3-pyrroline-1-carboxylate, also known as N-benzyloxycarbonyl 3-pyrroline, is an N-substituted 3-pyrroline. Its enthalpy of vaporization at boiling point has been evaluated. Its epoxidation using m-chloroperbenzoic acid (m-CPBA) has been reported.

application

Benzyl 3-pyrroline-1-carboxylate (N-Benzyloxycarbonyl 3-pyrroline) may be used to prepare (+)(3R,4R)-3,4-dihydroxy pyrrolidine via enantioselective trans-dihydroxylation catalyzed by frozen/thawed cells of Sphingomonas sp. HXN-200.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 598-598 (2014)
Asymmetric trans-dihydroxylation of cyclic olefins by enzymatic or chemo-enzymatic sequential epoxidation and hydrolysis in one-pot.
Xu Y, et al.
Green Chemistry, 13(9), 2452-2458 (2011)
Dongliang Chang et al.
The Journal of organic chemistry, 68(22), 8599-8606 (2003-10-25)
The bacterial strain Sphingomonas sp. HXN-200 was used to catalyze the trans dihydroxylation ofN-substituted 1,2,5,6-tetrahydropyridines 1 and 3-pyrrolines 4 giving the corresponding 3,4-dihydroxypiperidines 3 and 3,4-dihydroxypyrrolidines 6, respectively, with high enantioselectivity and high activity. The trans dihydroxylation was sequentially catalyzed
David R Williams et al.
Journal of the American Chemical Society, 126(11), 3434-3435 (2004-03-18)
A powerful cascade reaction process for the construction of functionalized cis-bicyclo[3.3.0]octenes has been developed. Carbolithiation of 3-methylene-1,4-cyclooctadiene with 1 degrees , 2 degrees , or 3 degrees alkyllithium reagents leads to cyclooctadienyl anions, which undergo disrotatory electrocyclization and subsequent trapping
S L Gwaltney et al.
Bioorganic & medicinal chemistry letters, 11(7), 871-874 (2001-04-11)
Sulfonate analogues of combretastatin A-4 have been prepared. These compounds compete with colchicine and combretastatin A-4 for the colchicine binding site on tubulin and are potent inhibitors of tubulin polymerization and cell proliferation. Importantly, these compounds also inhibit the proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service